Skip to Content
MilliporeSigma
All Photos(1)

Documents

6.18752

Supelco

Carbon Disulphide

Synonym(s):

Carbon disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
Beilstein:
1098293
MDL number:
UNSPSC Code:
12191502

grade

ACS reagent

vapor density

2.67 (vs air)

autoignition temp.

212 °F

expl. lim.

50 %

refractive index

n20/D 1.627 (lit.)

bp

46 °C (lit.)

mp

−112-−111 °C (lit.)

density

1.266 g/mL at 25 °C (lit.)

SMILES string

S=C=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Facile synthesis of tetrahydroquinoline containing dithiocarbamate derivatives via one-pot sequential multicomponent reaction.: This research discusses a simplified method for synthesizing tetrahydroquinoline derivatives with dithiocarbamate, a process involving Carbon Disulphide, with applications in developing new chemical entities (Senapati et al., 2024).


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service