Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.94513

Supelco

Carbon disulfide

EMPLURA®

Synonym(s):

Carbon disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
MDL number:
UNSPSC Code:
12352100

product line

EMPLURA®

storage temp.

room temp

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Target Organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service