Skip to Content
MilliporeSigma
All Photos(1)

Documents

CX0397

Supelco

Carbon Disulfide

low benzene OmniSolv®

Synonym(s):

Carbon Disulfide, Carbon Bisulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
MDL number:
UNSPSC Code:
12191602
EC Index Number:
200-843-6

grade

for use as solvent

Quality Level

vapor pressure

398 hPa ( 20 °C)

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

technique(s)

HPLC: suitable

impurities

≤0.03% Water (H2O)
≤200 ppb Benzene (CH6)

evapn. residue

≤3 ppm

color

APHA: ≤10

bp

46.5 °C/1013 hPa

mp

-111.6 °C

transition temp

flash point -30 °C

solubility

2.1 g/L

density

1.26 g/cm3 at 20 °C

UV cutoff

≤382 nm

UV absorption

λ: 382 nm Amax: ≤1.00
λ: 388 nm Amax: ≤0.50
λ: 403 nm Amax: ≤0.10
λ: 410 nm Amax: ≤0.05
λ: 500 nm Amax: ≤0.01

application(s)

general analytical

shipped in

ambient

storage temp.

room temp

SMILES string

C(=S)=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Related Categories

Application

CX0397-6 is packaged in poly coated glass bottles
low benzene OmniSolv®

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.9% minBenzene (C6H6)200ppb maxColor (APHA)10 maxEvaporation residue3ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardUV Abs. at 382 nm1.00AU maxUV Abs. at 388 nm0.50AU maxUV Abs. at 403 nm0.10AU maxUV Abs. at 410 nm0.05AU maxUV Abs. at 500 nm0.01AU maxUV Cut-off382nm maxWater (H2O)0.03% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Target Organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service