Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.31465

Sigma-Aldrich

Val-boroPro

≥98% (HPLC), solid, DPP-IV, DPP8 and DPP9 inhibitor, Calbiochem®

Synonym(s):

Val-boroPro, PT-100, Talabostat, H₂N-( S)-Val-( R)-boroPro-OH, CH₃SO₃H, H-( S)-Val-( R)-boroPro-OH, CH₃SO₃H, Valinyl-L-boroproline, CH₃SO₃H, DPP Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H23BN2O6S
CAS Number:
Molecular Weight:
310.18
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Val-boroPro,

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

180 pM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

light beige

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

B(O)(O)[C@H]1N(CCC1)C(=O)[C@@H]([N+H3])C(C)C.[S](=O)(=O)([O-])C

InChI

1S/C9H19BN2O3.CH4O3S/c1-6(2)8(11)9(13)12-5-3-4-7(12)10(14)15;1-5(2,3)4/h6-8,14-15H,3-5,11H2,1-2H3;1H3,(H,2,3,4)/t7-,8-;/m0./s1

InChI key

OXYYOEIGQRXGPI-WSZWBAFRSA-N

General description

A prolineboronic acid-based broad-spectrum DDP IV activity and/or structural homologue (DASH) family dipeptidyl peptidases inhibitor that converts in solution between the protonated active form and a cyclized inactive form due to a reversible intramolecular B-N dative bond formation of the non-protonated Val-boroPro (kinact = 6.4 x 10--4 s- and kcyc = 6.4 x 10-5 s- at pH 7.8). Although equilibrium favors cyclization at basic pH, high affinity binding (kon & koff toward CD26 at pH 7.8 = 5.02 x 106 M-s- and 77.8 x 10-6 s-, respectively) drives the equilibrium away from cyclization and traps the active form in enzyme-bound state with prolonged incubation. Reported to inhibit DPP7/DPPII/QPP (IC50/[Substrate]/rxn time at pH 5.5 = 15 nM/500 µM Lys-Pro-MNA/1 h & 310 nM/50 µM Nle-Pro-AMC/15 min), CD26/DPPIV (IC50 = 26 nM; 500 µM Ala-Pro-MNA for 1 h at pH 7.8), fibroblast activation protein/FAP (IC50 = 40 nM; 250 µM Ala-Pro-AFC at pH 2.0), DPP8 (IC50 = 4 nM; 100 µM Ala-Pro-AFC for 15 min at pH 8.0), DDP9 (IC50 = 11 nM; 100 µM Gly-Pro-AMC for 30 min at pH 7.4) with little potency against 9 other proteases (IC50 >100 µM; aminopeptidase P, chymoptrypsin, leukocyte elastase, plasma kallikrein, plasmin, prolidase, thrombin, trypsin, tryptase), while contradictory reports regarding prolyl endopeptidase/PEP/PREP inhibition exist (e.g. IC50 = 390 nM/50 µM Z-Gly-Pro-AMC/pH 7.5/30 min vs. 25 µM/500 µM Z-Gly-Pro-MNA/pH 7.8/60 min). Shown to promote the propagation of human BM CD34+ cells by stimulating stromal cells cytokine production in vitro (min EC 0.1 nM) and display hematopoietic stimulatory effect in mice in vivo (min ED 2 µg/mouse/12 h; p.o.). Although orally available in mice and rats, caution must be taken not to exceed toxicity levels (MTD = 25 µg/mL using Sprague-Dawley rats; LD50 ~0.5 mg/mL in 24 survival test using Fisher rats; p.o.) for in vivo studies.
An orally available amino boronic acid dipeptide with antihyperglycemic and anticancer properties. Acts as a highly potent, reversible, and transition state analog inhibitor of dipeptidyl peptidase IV (DPP-IV; IC50 = 26 nM; Ki = 180 pM). Also inhibits the activity of other prolyl peptidases (IC50 = 15 nM for DPP-II; and Ki = 1.5 nM and 760 pM for DPP8 and DPP9, respectively) and at much higher concentrations affects the activities of quiescent cell proline dipeptidase (QPP, DPP7; IC50 = 310 nM), PEP (IC50 = 390 nM) and fibroblast activation protein (FAP; IC50 = 560 nM). However, it does not inhibit the activity of trypsin, chymoptrypsin, thrombin, plasmin, tryptase, leukocyte elastase, and plasma kallikrein even at high levels (~100 µM). Promotes hematopoiesis and the growth of primitive hematopoietic progenitor cells by increasing the production of G-CSF, IL-6, and IL-11 by bone marrow stromal cells in mice. Although it has no direct cytotoxic effect on tumors in vitro, but oral administration in mice is shown to reduce the growth of syngeneic tumors derived from fibrosarcoma (WEHI164), lymphoma (EL4 and A20/2J), melanoma (B16-F10), and mastocytoma (P815) cell lines and augments antibody-dependent cell-mediated cytotoxicity. Note: In aqueous physiological solutions, it can cyclize in a time-dependent manner causing a 100-fold diminution in its inhibitory potency. However, at lower pH range it remains in an active open chain form. Maximum tolerated dose in mice or rats is ~ 25 µg/kg.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
DPP-IV, DPP8 and DPP9
Reversible: yes
Secondary Target
QPP, PEP and FAP
Target IC50: 15 nM for DPP-II

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Poplawski, S.E., et al. 2011, J. Med. Chem.54, 2022.
Connolly, B.A., et al. 2008. J. Med. Chem.51, 6005.
Lankas, G.R., et al. 2005. Diabetes54, 2988.

Cheng, J.D., et al. 2005. Mol. Cancer Ther.4, 351.
Adams, S., et al. 2004. Cancer Res.64, 5471.
Jones, B., et al. 2003. Blood102, 1641.
Coutts, S.J., et al. 1996. J. Med. Chem.39, 2087.
Snow, R.J., et al. 1994. J. Am. Chem. Soc.116, 10860.
Kelly, T.A., et al. 1993. J. Am. Chem. Soc.115, 12637.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan D Cheng et al.
Molecular cancer therapeutics, 4(3), 351-360 (2005-03-16)
Tumor-associated fibroblasts are functionally and phenotypically distinct from normal fibroblasts that are not in the tumor microenvironment. Fibroblast activation protein is a 95 kDa cell surface glycoprotein expressed by tumor stromal fibroblasts, and has been shown to have dipeptidyl peptidase
Sharlene Adams et al.
Cancer research, 64(15), 5471-5480 (2004-08-04)
The amino boronic dipeptide, PT-100 (Val-boro-Pro), a dipeptidyl peptidase (DPP) inhibitor, has been shown to up-regulate gene expression of certain cytokines in hematopoietic tissue via a high-affinity interaction, which appears to involve fibroblast activation protein. Because fibroblast activation protein is
George R Lankas et al.
Diabetes, 54(10), 2988-2994 (2005-09-28)
Dipeptidyl peptidase (DPP)-IV inhibitors are a new approach to the treatment of type 2 diabetes. DPP-IV is a member of a family of serine peptidases that includes quiescent cell proline dipeptidase (QPP), DPP8, and DPP9; DPP-IV is a key regulator
S J Coutts et al.
Journal of medicinal chemistry, 39(10), 2087-2094 (1996-05-10)
A series of prolineboronic acid (boroPro) containing dipeptides were synthesized and assayed for their ability to inhibit the serine protease dipeptidyl peptidase IV (DPPIV). Inhibitory activity, which requires the (R)-stereoisomer of boroPro in the P1 position, appears to tolerate a
Beth A Connolly et al.
Journal of medicinal chemistry, 51(19), 6005-6013 (2008-09-12)
Dipeptidyl peptidase IV (DPP-IV; E.C. 3.4.14.5), a serine protease that degrades the incretin hormones GLP-1 and GIP, is now a validated target for the treatment of type 2 diabetes. Dipeptide boronic acids, among the first, and still among the most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service