MilliporeSigma
  • PT-100, a small molecule dipeptidyl peptidase inhibitor, has potent antitumor effects and augments antibody-mediated cytotoxicity via a novel immune mechanism.
PT-100, a small molecule dipeptidyl peptidase inhibitor, has potent antitumor effects and augments antibody-mediated cytotoxicity via a novel immune mechanism.
Cancer research (2004-08-04)
Sharlene Adams, Glenn T Miller, Michael I Jesson, Takeshi Watanabe, Barry Jones, Barbara P Wallner
ABSTRACT
The amino boronic dipeptide, PT-100 (Val-boro-Pro), a dipeptidyl peptidase (DPP) inhibitor, has been shown to up-regulate gene expression of certain cytokines in hematopoietic tissue via a high-affinity interaction, which appears to involve fibroblast activation protein. Because fibroblast activation protein is also expressed in stroma of lymphoid tissue and tumors, the effect of PT-100 on tumor growth was studied in mice in vivo. PT-100 has no direct cytotoxic effect on tumors in vitro. Oral administration of PT-100 to mice slowed growth of syngeneic tumors derived from fibrosarcoma, lymphoma, melanoma, and mastocytoma cell lines. In WEHI 164 fibrosarcoma and EL4 and A20/2J lymphoma models, PT-100 caused regression and rejection of tumors. The antitumor effect appeared to involve tumor-specific CTL and protective immunological memory. PT-100 treatment of WEHI 164-inoculated mice increased mRNA expression of cytokines and chemokines known to promote T-cell priming and chemoattraction of T cells and innate effector cells. The role of innate activity was further implicated by observation of significant, although reduced, inhibition of WEHI 164 and A20/2J tumors in immunodeficient mice. PT-100 also demonstrated ability to augment antitumor activity of rituximab and trastuzumab in xenograft models of human CD20(+) B-cell lymphoma and HER-2(+) colon carcinoma where antibody-dependent cytotoxicity can be mediated by innate effector cells responsive to the cytokines and chemokines up-regulated by PT-100. Although CD26/DPP-IV is a potential target for PT-100 in the immune system, it appeared not to be involved because antitumor activity and stimulation of cytokine and chemokine production was undiminished in CD26(-/-) mice.
Sigma-Aldrich
Val-boroPro