MilliporeSigma
  • Dipeptide boronic acid inhibitors of dipeptidyl peptidase IV: determinants of potency and in vivo efficacy and safety.
Dipeptide boronic acid inhibitors of dipeptidyl peptidase IV: determinants of potency and in vivo efficacy and safety.
Journal of medicinal chemistry (2008-09-12)
Beth A Connolly, David G Sanford, Amrita K Chiluwal, Sarah E Healey, Diane E Peters, Matthew T Dimare, Wengen Wu, Yuxin Liu, Hlaing Maw, Yuhong Zhou, Youhua Li, Zhiping Jin, James L Sudmeier, Jack H Lai, William W Bachovchin
ABSTRACT
Dipeptidyl peptidase IV (DPP-IV; E.C. 3.4.14.5), a serine protease that degrades the incretin hormones GLP-1 and GIP, is now a validated target for the treatment of type 2 diabetes. Dipeptide boronic acids, among the first, and still among the most potent DPP-IV inhibitors known, suffer from a concern over their safety. Here we evaluate the potency, in vivo efficacy, and safety of a selected set of these inhibitors. The adverse effects induced by boronic acid-based DPP-IV inhibitors are essentially limited to what has been observed previously for non-boronic acid inhibitors and attributed to cross-reactivity with DPP8/9. While consistent with the DPP8/9 hypothesis, they are also consistent with cross-reactivity with some other intracellular target. The results further show that the potency of simple dipeptide boronic acid-based inhibitors can be combined with selectivity against DPP8/9 in vivo to produce agents with a relatively wide therapeutic index (>500) in rodents.
Sigma-Aldrich
Val-boroPro