Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

238407

Sigma-Aldrich

Creatine Phosphokinase-MM Isoenzyme, Human Skeletal Muscle

Creatine Phosphokinase-MM Isoenzyme, Human Skeletal Muscle, is a native creatine phosphokinase-MM f that catalyzes the conversion of creatine and ATP to phosphocreatine and ADP.

Synonym(s):

hCPK-MM

Sign Into View Organizational & Contract Pricing

Select a Size

50 I.U.
$258.00

$258.00


Please contact Customer Service for Availability


Select a Size

Change View
50 I.U.
$258.00

About This Item

EC Number:
UNSPSC Code:
12352202
NACRES:
NA.54

$258.00


Please contact Customer Service for Availability

Quality Level

form

liquid

specific activity

≥300 units/mL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−20°C

General description

Native creatine phosphokinase-MM from human skeletal muscle. Catalyzes the reversible conversion of creatine and ATP to phosphocreatine and ADP. Plasma levels of CPK-MM are a useful indicator of acute myocardial infarction.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will catalyze the formation of 1 µmol of ATP from ADP and creatine phosphate per min at 37°C, pH 7.4.

Physical form

In 10 mM Sodium Chloride, 5 mM Succinate, 5 mM β-Mercaptoethanol, 1 mM EDTA, 50% Glycerol, pH 7.1 +/- 0.1.

Preparation Note

Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Baral, R., et al. 1993. Intern. Med.33, 210.
Wu, A.H.B. 1992. Lab. Med.23, 303.
Bergmeyer, H.U. 1974. Methods Enzymatic Anal.2, 789.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service