Skip to Content
MilliporeSigma
All Photos(1)

Documents

238395

Sigma-Aldrich

Creatine Phosphokinase, Porcine Heart

Creatine Phosphokinase, Porcine Heart, CAS 9001-15-4, is a native creatine phosphokinase that catalyzes the reversible conversion of creatine and ATP to creatine phosphate and ADP.

Synonym(s):

Creatine Phosphokinase, Porcine Heart, pCPK

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

Quality Level

Assay

≥98% (UV, TLC)

form

lyophilized

specific activity

≥300 U/mg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

aqueous buffer: soluble
water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Native creatine phosphokinase from porcine heart. Catalyzes the reversible conversion of creatine and ATP to creatine phosphate and ADP. Often used as a component of ATP-generating systems.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will transfer 1.0 µmol phosphate from creatine phosphate to ADP per min at 37°C, pH 7.4.

Reconstitution

Following reconstitution store in the refrigerator (4°C). Add 0.1% BSA to solutions prior to storage. Stock solutions are stable for up to 1 week at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service