Skip to Content
MilliporeSigma
All Photos(1)

Documents

2384

Sigma-Aldrich

Creatine Phosphokinase, Rabbit Skeletal Muscle

Synonym(s):

Creatine Phosphokinase, Rabbit Skeletal Muscle, rCPK

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.21

biological source

rabbit skeletal muscle

Quality Level

form

lyophilized

specific activity

≥300 U/mg, 25 °C
750 U/mg, 37 °C

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

0.1 M imidazole buffer, pH 6.6: soluble
0.5% NaHCO3, pH 9.0: soluble

foreign activity

ATPase, hexokinase, myokinase <0.001%

shipped in

ambient

storage temp.

−20°C

General description

Native creatine phosphokinase from rabbit muscle. Catalyzes the conversion of creatine and ATP to creatine phosphate and ADP. Has an optimal pH of 9.0 for the forward reaction and 6.0-7.0 for the reverse reaction.
Native creatine phosphokinase from rabbit muscle. Catalyzes the reversible conversion of creatine and ATP to creatine phosphate and ADP. Has an optimal pH of 9.0 for the forward reaction and 6.0-7.0 for the reverse reaction.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will catalyze the conversion of 1.0 µmol creatine phosphate per min at 25°C or 37°C, pH 6.8.

Reconstitution

Following reconstitution in 0.5% NaHCO₃, refrigerate (4°C). Do not freeze or shake, as SH groups may be oxidized and form linkages between protein molecules, resulting in precipitation, turbidity, and loss of activity. NaHCO₃ stock solutions are stable for 2-3 days at 4°C. Following reconstitution in 0.1 M imidazole buffer, aliquot and freeze (-20°C). Imidazole stock solutions are stable for up to 4 weeks at -20°C. Avoid freeze/thaw cycles, as 20-30% of activity will be lost with each freeze/thaw cycle.

Other Notes

Bessman, S.P. 1987. Anal. Biochem.161, 519.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service