Skip to Content
MilliporeSigma
All Photos(1)

Documents

2150-OP

Millipore

Ammonium Sulfate

OmniPur® Grade, >= 99.5%, Molecular biology grade.

Synonym(s):

Ammonium sulfate, Diammonium Sulfate, Sulfuric Acid Sodium Salt, Sulphuric Acid Sodium Salt, Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
MDL number:
UNSPSC Code:
12352203
NACRES:
NA.28

Quality Level

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

mp

>280 °C (dec.) (lit.)

density

1.77 g/mL at 25 °C (lit.)

foreign activity

DNAse, none detected
Phosphatase, none detected
Protease, none detected
RNAse, none detected

shipped in

ambient

storage temp.

10-30°C

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Molecular biology grade.

Specifications:
A (260 nm, 0.1 M, water): ≤0.01AU
A (280 nm, 0.1 M, water): ≤0.01AU
Assay: ≥99.5%
DNase: None detected
Heavy metals (as Pb): ≤0.0002%
Phosphatase: None detected
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Isolation and identification of milk-clotting proteases from Prinsepia utilis Royle and its application in cheese processing.: This research highlights the use of ammonium sulfate in precipitating specific enzymes during cheese production, optimizing the isolation of milk-clotting proteases which are crucial for improving cheese yield and quality (Zhang L et al., 2024).

  • Diffusion-driven fed-batch fermentation in perforated ring flasks.: Demonstrates an innovative application of ammonium sulfate in fermentation processes, where it serves as a nitrogen source to sustain microbial growth and enhance biotechnological production efficiencies (Lüchtrath C et al., 2024).

  • Extraction of Protein and Bioactive Compounds from Mediterranean Red Algae (Sphaerococcus coronopifolius and Gelidium spinosum) Using Various Innovative Pretreatment Strategies.: This study utilizes ammonium sulfate in a salting-out process to efficiently extract valuable proteins and bioactive compounds from red algae, contributing to advancements in nutritional supplements and pharmaceuticals (Dhaouafi J et al., 2024).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service