Skip to Content
MilliporeSigma
All Photos(1)

Documents

168355

Millipore

Ammonium sulfate

Synonym(s):

(NH₄)₂SO₄, Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
MDL number:
UNSPSC Code:
12352314
NACRES:
NA.25

Assay

>99% (formol method)

Quality Level

form

crystals (Fine)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

mp

>280 °C (dec.) (lit.)

solubility

water: 1 M

density

1.77 g/mL at 25 °C (lit.)

cation traces

heavy metals: <5 ppm

shipped in

ambient

storage temp.

10-30°C

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A 5% aqueous solution has a pH of 5.0-6.0. Commonly used for protein precipitation/purification.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature. Stock solutions are stable for up to 1 year at room temperature.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service