1.28218
Ammonium sulfate
EMPROVE® EXPERT, ChP, NF, ACS
Pharma Manufacturing
Synonym(s):
Mascagnite
Select a Size
Select a Size
About This Item
Recommended Products
grade
ACS reagent
Quality Level
Agency
ChP
NF
vapor pressure
<1 Pa ( 25 °C)
product line
EMPROVE® EXPERT
form
solid
potency
2840 mg/kg LD50, oral (rat)
pH
5 (100 g/L in H2O)
mp
>280 °C (dec.) (lit.)
solubility
754-764 g/L
density
1.77 g/mL at 25 °C (lit.)
bulk density
850 kg/m3
application(s)
liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical
storage temp.
2-30°C
SMILES string
N.N.OS(O)(=O)=O
InChI
1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)
InChI key
BFNBIHQBYMNNAN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Due to its low microbial and endotoxin limits, Ammonium sulfate Emprove® Expert is especially suitable for high-risk applications.
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service