Skip to Content
MilliporeSigma

137119

SAFC

Phospho-L-Tyrosine Disodium Salt

EMPROVE® EXPERT

Pharma Manufacturing

Synonym(s):

Phospho-L-tyrosine disodium salt, PTyr, Phospho-Tyrosine, Phosphotyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10NNa2O6P
CAS Number:
Molecular Weight:
305.13
MDL number:
UNSPSC Code:
12352209
Pricing and availability is not currently available. Contact Technical Service

biological source

synthetic

Quality Level

product line

EMPROVE® EXPERT

form

powder or crystals

packaging

pkg of 1 kg (in PE drum, 2 inner PE liners 1371191000)
pkg of 10 kg (in PE drum, 2 inner PE liners 1371199010)
pkg of 100 g (in PE tub, inner glas bottle 1371190100)

pH

5.5-9.0 (10 g/L in H2O)

solubility

25 g/L
≤70 g/L (in complex feed)

suitability

suitable for manufacturing use (cell culture)

storage temp.

2-8°C

SMILES string

[Na].[Na].[P](=O)(Oc1ccc(cc1)C[C@H](N)C(=O)O)(O)O

InChI

1S/C9H12NO6P.2Na/c10-8(9(11)12)5-6-1-3-7(4-2-6)16-17(13,14)15;;/h1-4,8H,5,10H2,(H,11,12)(H2,13,14,15);;/t8-;;/m0../s1

InChI key

JRFJYPIBECDABY-JZGIKJSDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Modified amino acids are inhouse manufactured amino acid derivatives with specific properties enabling the intensification of cell culture processes.

Together with the companion product Sulfo-Cysteine sodium salt, the new modified amino acid Phospho-Tyrosine disodium salt can be used as replacement for tyrosine to generate highly concentrated, neutral pH feeds. Both modified amino acids eliminate the need for alkaline feeds, which are normally applied to ensure solubility and stability of the unmodified amino acids tyrosine and cysteine.
Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Features and Benefits

  • Reduced complexity in fed-batch process
  • High concentrations of modified tyrosine in main feeds at neutral pH
  • Enhanced solubility up to 70g/l in complex feed
  • Prevention of caustic shocks in the bioreactor due to high pH feeds
  • More convenient preparation process with less contamination risks
  • Higher feed stability at room temperature

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service