Skip to Content
MilliporeSigma

1.37219

SAFC

Phospho-L-Tyrosine Disodium Salt

EMPROVE® EVOLVE

Pharma Manufacturing

Synonym(s):

Phospho-L-tyrosine disodium salt, PTyr, Phospho-Tyrosine, Phosphotyrosine

Sign Into View Organizational & Contract Pricing

Select a Size

1372190100
$1,134.00
1372191000
$6,747.00
1372199010
$55,639.00

$1,134.00


Please contact Customer Service for Availability


Select a Size

Change View
1372190100
$1,134.00
1372191000
$6,747.00
1372199010
$55,639.00

About This Item

Empirical Formula (Hill Notation):
C9H10NNa2O6P
CAS Number:
Molecular Weight:
305.13
MDL number:
UNSPSC Code:
12352209

$1,134.00


Please contact Customer Service for Availability

product line

EMPROVE® EVOLVE

Quality Level

application(s)

pharma/biopharma processes

storage temp.

2-8°C

SMILES string

[Na].[Na].[P](=O)(Oc1ccc(cc1)C[C@H](N)C(=O)O)(O)O

InChI

1S/C9H12NO6P.2Na/c10-8(9(11)12)5-6-1-3-7(4-2-6)16-17(13,14)15;;/h1-4,8H,5,10H2,(H,11,12)(H2,13,14,15);;/t8-;;/m0../s1

InChI key

JRFJYPIBECDABY-JZGIKJSDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

Application

Modified amino acids are inhouse manufactured amino acid derivatives with specific properties enabling the intensification of cell culture processes.
Together with the companion product Sulfo-Cysteine sodium salt, the new modified amino acid Phospho-Tyrosine disodium salt can be used as replacement for tyrosine to generate highly concentrated, neutral pH feeds. Both modified amino acids eliminate the need for alkaline feeds, which are normally applied to ensure solubility and stability of the unmodified amino acids tyrosine and cysteine.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service