Skip to Content
MilliporeSigma

137116

SAFC

L-Cysteine S-sulfate sodium salt sesquihydrate

EMPROVE® EXPERT

Pharma Manufacturing

Synonym(s):

L-Cysteine S-sulfate sodium salt sesquihydrate, S-Sulfocysteine, SCys, Sulfo-Cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6NNaO5S2 · 1.5H2O
CAS Number:
Molecular Weight:
250.23
UNSPSC Code:
12352209

biological source

synthetic

Quality Level

product line

EMPROVE® EXPERT

form

powder or crystals

packaging

pkg of 1 kg (1371161000)
pkg of 100 g (1371160100)
pkg of 5 kg (1371165000)

solubility

water: soluble (Up to 1.3M (room temp))

suitability

suitable for manufacturing use (cell culture media)

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Modified amino acids are inhouse manufactured amino acid derivatives with specific properties enabling the intensification of cell culture processes.

Together with the companion product Phospho-L-Tyrosine disodium Salt disodium salt, the new modified amino acid S-Sulfocysteine sodium salt can be used as replacement for cysteine to generate highly concentrated, neutral pH feeds. Both modified amino acids eliminate the need for alkaline feeds, which are normally applied to ensure solubility and stability of the unmodified amino acids tyrosine and cysteine.
Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Features and Benefits

  • Reduced complexity in fed-batch process
  • High concentrations of modified cysteine in main feeds at neutral pH
  • Prevention of caustic shocks in the bioreactor due to high pH feeds
  • More convenient preparation process with less contamination risks
  • Reduction of recombinant protein fragmentation and decrease in trisulfide bond content
  • Higher feed stability at room temperature

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service