Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.25054

Millipore

Fractogel® EMD TMAE (M)

MiniChrom Column Fractogel® TMAE (M), volume 1 mL, prepacked column

Synonym(s):

MiniChrom Column Fractogel® TMAE (M), 1 ml

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$253.00

$253.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
$253.00

About This Item

UNSPSC Code:
41115711

$253.00


Please contact Customer Service for Availability

ligand

(Trimethylammoniumethyl)

material

polypropylene housing

product line

Fractogel®

form

prepacked column

parameter

≤8 bar pressure
240 cm/hr max. flow rate

column I.D. × L

8 mm × 20 mm

volume

1 mL

matrix active group

Methacrylate

mean particle size

40-90 μm

capacity

~100 mg, BSA binding capacity

application(s)

gene therapy
mAb
recombinant protein

separation technique

anion exchange

storage temp.

15-25°C

General description

8 x 20 mm

Features and Benefits

Fractogel® EMD TMAE (M) enables:

  • High protein loading conditions resulting in good selectivity and resolution with the tentacle technology
  • Excellent binding to large viruses at increased loading
  • Homogenous binding with high selectivity and purity
  • Reduce operating costs and resin replacement with enduring durability - allowing for multiple cycles of column regeneration and sanitization
  • Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service