1.25061
Fractogel® EMD SO3 ⁻ (M) Resin
MiniChrom Column Fractogel® SO3 (M), volume 1 mL, prepacked column
Synonym(s):
MiniChrom Column Fractogel® SO3 (M), 1 ml
Select a Size
$253.00
Select a Size
About This Item
$253.00
Recommended Products
ligand
(Sulfoisobutyl)
Quality Level
material
polypropylene housing
product line
Fractogel®
form
prepacked column
parameter
≤8 bar pressure
~200 cm/hr max. flow rate
column I.D. × L
8 mm × 20 mm
volume
1 mL
matrix active group
Methacrylate
mean particle size
40-90 μm
capacity
~130 mL, resin binding capacity
application(s)
mAb
recombinant protein
vaccine development
separation technique
cation exchange
storage temp.
2-30°C
General description
Features and Benefits
- Increased binding strength at low pH
- High aggregate removal efficiency combined with high dynamic binding capacity
- Lower elution volumes for the highest purity levels
- Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution
Due to the titration behavior, the ion exchange capacity can be used from pH 2 to pH 12. The separation of proteins is based on reversible electrostatic interactions between the surface′s positively charged regions and the support. Proteins are retained efficiently on Fractogel® EMD SO3- (M) when the pH of the buffer is about 1 unit below their isoelectric points (pl).
The strength of the binding depends on the following:
- the buffer system
- pH value of the buffer which determines the surface charge of the protein
- the concentration of the counter ions
- the charge density on the support (protein binding capacity)
Analysis Note
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service