Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

125057

Millipore

Fractogel® EMD TMAE (S)

MiniChrom Column Fractogel® TMAE (S), volume 1 mL, prepacked column

Synonym(s):

MiniChrom Column Fractogel® TMAE (S), 1 ml

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$253.00

$253.00


Please contact Customer Service for Availability


Select a Size

Change View
1 UNIT
$253.00

About This Item

UNSPSC Code:
41115711

$253.00


Please contact Customer Service for Availability

ligand

(Trimethylammoniumethyl)

material

polypropylene housing

product line

Fractogel®

form

prepacked column

parameter

≤8 bar pressure
~80 cm/hr max. flow rate

column I.D. × L

8 mm × 20 mm

volume

1 mL

matrix active group

Methacrylate

mean particle size

20-40 μm

capacity

~100 mg, BSA binding capacity

application(s)

gene therapy
recombinant protein
vaccine development

shipped in

ambient

storage temp.

15-25°C

General description

8 x 20mm

Analysis Note

Appearance: passes test

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore strategies for separating empty and filled capsids in viral vector production. Learn about upstream strategies and how trends in AAV capsid design impact purification.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service