Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.16971

Supelco

Nitrate Test Strips, reflectometric

5-225 mg/L (NO₃⁻), REFLECTOQUANT®

Synonym(s):

Nitrate chemical test strips, Nitrate test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Nitrate Test, reflectometric, 5-225 mg/L (NO3-), for use with REFLECTOQUANT®

description

quantitative

Quality Level

specific analyte(s)

nitrate

reaction suitability

reaction type: Griess reaction

measuring range

5-225 mg/L (NO3-)

compatibility

RQflex® 10 Plus
RQflex® 10
RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips 5 - 225 mg/l NO₃⁻ Reflectoquant®
The Reflectoquant Nitrate test strips can be used for the rapid determination of the nitrate content in water and also with fruit and vegetables to help decide whether a given delivery batch can be released for use in a specific product (e.g. baby foods). On the strip nitrate ions are reduced to nitrite ions by a reducing agent. In the presence of an acidic buffer, these nitrite ions react with an aromatic amine to form a diazonium salt, which in turn reacts with N-(1naphthyl)-ethylene-diamine to form a red-violet azo dye that is determined reflectometrically on the RQflex. For pre-testing a sample in an economic and quick way to check for nitrite interferences or to estimate the nitrate concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for nitrate and nitrite. Further quantitative determination methods can be found with our Spectroquant tests.

Linkage

Replaces: 16971; 16971-1

Analysis Note

function control: conforms
Assay
- standard 1:
22 - 28 mg/l
- standard 2: 90 - 110 mg/l
- standard 3: 180 - 220 mg/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Reflectometric nitrate determination in vegetables using Griess reagent and Reflectoquant® system.

Reflectometric determination of nitrate in wastewater using Griess reagent and Reflectoquant® test strips.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service