Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.03981

Sigma-Aldrich

M-FIX® spray fixative

for cytodiagnosis

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$33.20

$33.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$33.20

About This Item

UNSPSC Code:
12352401
NACRES:
NA.21

$33.20


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

liquid

IVD

for in vitro diagnostic use

color

colorless

kinematic viscosity

2.86 cSt(20 °C)

transition temp

flash point 12 °C

density

0.825 g/cm3 at 20 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

M-FIX® spray fixative - for cytodiagnosis, is a fixative spray that is suitable for the wet fixation of gynaecological and non-gynaecological preparations processed in Papanicolaou staining. M-FIX® contains polyglycols in alcoholic solution and a rapid and secure fixation of specimens is possible. Its formulation enables stabilization of the material upon alcohol fixation and forms a protective film against desiccation and damage to the specimen material.
100 ml M-FIX® is sufficient for approx. 250 applications. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Legal Information

M-FIX is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service