Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.07960

Sigma-Aldrich

Entellan

rapid mounting medium for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401
NACRES:
NA.21

Quality Level

autoignition temp.

>250 °C

expl. lim.

1.2-8 % (v/v) toluene()

IVD

for in vitro diagnostic use

color

colorless

refractive index

n20/D 1.492-1.500

transition temp

flash point 8 °C

density

0.93 g/cm3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Entellan- rapid mounting medium for microscopy, is a ready-to-use, water-free mounting medium for microscopy for the permanent mounting of specimen is an anhydrous mounting medium for microscopy used for the permanent mounting and storage of specimens, and consists of a polymer made of mixed acrylates dissolved in toluene. As it contains toluene, it should be used with water-free specimens that have been processed with xylene previous mounting. Due to its registration as IVD product and CE certification, it can be used in the manual or automated application in a diagnostic laboratory. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Refractive index (20°C) 1.492 - 1.500 at 20°C
Density (20°C / 4°C) 0.925 - 0.935 g/cm3
Viscosity (20°C) 60 - 100 mPa*s
Fluorescence < 100 ppb

Analysis Note

Refractive index (n 20/D): 1.492 - 1.500
Density (d 20 °C/ 4 °C): 0.925 - 0.935
Fluorescence (as quinine at 365 nm): ≤ 100 ppb
Viscosity (20 °C): 60 - 100 mPa∙s
Suitability for microscopy: passes test

Legal Information

ENTELLAN is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F

Flash Point(C)

8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meike M Rogalla et al.
eNeuro, 7(3) (2020-04-25)
Alterations in inhibitory circuits of the primary auditory cortex (pAC) have been shown to be an aspect of aging and age-related hearing loss (AHL). Several studies reported a decline in parvalbumin (PV) immunoreactivity in aged rodent pAC of animals displaying
Danica Sims et al.
Cancers, 12(2) (2020-02-27)
Rhabdomyosarcoma is a highly aggressive malignant cancer that arises from skeletal muscle progenitor cells and is the third most common solid tumour in children. Despite significant advances, rhabdomyosarcoma still presents a therapeutic challenge, and while targeted therapy has shown promise

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service