Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.00869

Sigma-Aldrich

Entellan new for cover slipper

for microscopy

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$276.00

$276.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
$276.00

About This Item

UNSPSC Code:
12352401
NACRES:
NA.21

$276.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

<8 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

>250 °C

expl. lim.

1.1-7.0 % (v/v) xylene()

IVD

for in vitro diagnostic use

color

colorless

refractive index

n/D 1.490-1.500

viscosity

500-600 mPa.s(20 °C)

bp

137-143 °C/1013 hPa

transition temp

flash point 23 °C

density

0.95 g/cm3 at 20 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Entellan new for cover slipper for microscopy, is a mounting medium for microscopy that is especially suited for standard commercial automated-mounting instruments that operate with glas coverslips. It is used as described in the instruction manual for cover slippers and the ideal amount of mounting agent is determined in a pilot run. There, empty cover glasses and specimen holders, according to the size of the cover glass and the size and thickness of the specimen, are used and these conditions are re-checked when a new bottle of the mounting medium is used. As its viscosity range is adjusted to a narrow range, the effort for new the calibration of the instrument is minimized. As the mounting medium is an IVD product and CE registered it can be used in diagnostics and for laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Refractive index (20°C) 1.490 - 1.500
Viscosity (20°C) 500 - 600 mPa*s

Analysis Note

Refractive index (n 20/D): 1.490 - 1.500
Fluorescence (as quinine at 365 nm): ≤ 250 ppb
Viscosity (20 °C): 500 - 600 mPa∙s
Suitability for microscopy: passes test

Legal Information

ENTELLAN is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service