Skip to Content
MilliporeSigma
All Photos(1)

Documents

700173P

Avanti

Dihydro T-MAS

powder

Synonym(s):

4,4-dimethyl-cholest-8(9)-en-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O
CAS Number:
Molecular Weight:
414.71
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

99% (TLC)

form

powder

packaging

package of 1 × 1 mg (700173P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@]12CCC([C@](CC[C@H](O)C3(C)C)(C)C3CC4)=C4C1CC[C@]2([H])[C@]([H])(C)CCCC(C)C

Biochem/physiol Actions

4,4-dimethyl-cholest-8(9)-en-3β-ol (Dihydro T-MAS) is a sterol involved in triggering the activation of meiosis in cultured oocytes of mice in vitro. It is an intermediate of the Kandutsch-Russell (K-R) pathway.

Packaging

5 mL Amber Glass Screw Cap Vial (700173P-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steroidal triterpenes of cholesterol synthesis
Avcimovivc J and Rozman D
Molecules (Basel), 18(4), 4002-4017 (2013)
Flux analysis of cholesterol biosynthesis in vivo reveals multiple tissue and cell-type specific pathways
Mitsche M A, et al.
eLife, 4, e07999-e07999 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service