Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.56057

Sigma-Aldrich

H-Pro-2-ClTrt resin

Novabiochem®

Synonym(s):

Chlorotrityl Resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

General description

Pre-loaded resin for synthesis of protected peptide acids containing a C-terminal proline amino-acid residue by Fmoc SPPS. Cleavage of the protected peptide is effected by treatment with 1% TFA in DCM or 20% TFE in DCM. The use of this resin enables the synthesis of peptides containing C-terminal Pro without risk of diketopiperazine formation.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Application

  • B Yakimova et al. Reported the use of H-Pro-2-ClTrt resin in the synthesis of short peptides with potential ACE inhibitory activity (B Yakimova, et al., 2021).
  • S Wang et al. Detailed the use of H-Pro-2-ClTrt resin in a study on aminoacyl chain translocation catalysed by a type II thioesterase domain in an unusual non-ribosomal peptide synthetase (S Wang, et al., 2022).
  • LP Vu et al. Described the use of H-Pro-2-ClTrt resin in the synthesis of inhibitors for serine proteases (LP Vu, et al., 2022).

Linkage

Replaces: 04-12-2805

Analysis Note

Color (visual): pale yellow to beige
Appearance of substance (visual): beads
Identity (of the substitution): passes test
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.30 - 1.10 mmol/g
Swelling Volume (in DMF): lot specific value
The polymer matrix is copoly (styrene - 1% DVB) 200 -400 mesh

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service