Skip to Content
MilliporeSigma
All Photos(2)

Documents

700073P

Avanti

T-MAS

Avanti Research - A Croda Brand

Synonym(s):

14-norlanosterol; 4,4-dimethylzymosterol; T-MAS; 110892, 4,4-dimethylcholest-8(9),24-dien-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H48O
CAS Number:
Molecular Weight:
412.69
UNSPSC Code:
12352211
NACRES:
NA.25

description

4,4-dimethyl-cholest-8(9),24-dien-3β-ol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700073P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−70°C

SMILES string

C[C@]12CCC([C@](CC[C@H](O)C3(C)C)(C)C3CC4)=C4C1CC[C@]2([H])[C@]([H])(C)CCC=C(C)C

InChI

1S/C29H48O/c1-19(2)9-8-10-20(3)22-12-13-23-21-11-14-25-27(4,5)26(30)16-18-29(25,7)24(21)15-17-28(22,23)6/h9,20,22-23,25-26,30H,8,10-18H2,1-7H3/t20-,22-,23+,25+,26+,28-,29-/m1/s1

InChI key

CHGIKSSZNBCNDW-QGBOJXOESA-N

General description

14-demethyl-lanosterol acts as an intermediate in cholesterol biosynthesis. It is used to stimulate oocytes for meiosis. 14-demethyl-lanosterol modulates cumulus expansion.

Application

14-demethyl-lanosterol has been used to reduce cancer cell viability and increase cancer cell sensitivity to erlotinib.

Packaging

5 mL Amber Glass Screw Cap Vial (700073P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The chemistry of the vitamins (1965)
Yen and Jaffe's Reproductive Endocrinology: Physiology, Pathophysiology, and Clinical Management (2009)
Targeting C4-demethylating genes in the cholesterol pathway sensitizes cancer cells to EGF receptor inhibitors via increased EGF receptor degradation
Sukhanova A, et al.
Cancer Discovery, 3(1), 96-111 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service