Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z290246

Supelco

Model 1000 oxygen trap

connections size 1/4 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NB.21

connections size

1/4 in.

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

High-pressure, nickel-plated steel cylinder filled with a high capacity oxygen adsorbent. Low initial cost and high capacity gives months of use and reduces down time. Exceptional efficiency typically results in less than 30 ppb oxygen at the outlet.

Oxygen traps protect the liquid phase in HPLC columns from oxidation, and are particularly useful for capillary columns.

Contains 500 g of adsorbent, capable of removing over 1 L of oxygen at standard temperature and pressure.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vikash Kumar et al.
Dalton transactions (Cambridge, England : 2003), 44(46), 19909-19917 (2015-10-31)
Cu(II) complexes of N2O2 type ligands, L(1)H2 and L(2)H2 [L(1)H2 = 6,6'-(((pyridin-2-ylmethyl)azanediyl)bis(methylene))bis(2,4-di-tert-butylphenol); L(2)H2 = 2,4-di-tert-butyl-6-(((3-(tert-butyl)-2-hydroxy-5-methylbenzyl)(pyridin-2-yl-methyl)amino)methyl)phenol], have been synthesized. Addition of nitrogen dioxide (NO2) in THF solutions of the complexes resulted in the nitration at the 4-position of a coordinated equatorial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service