MilliporeSigma
Copper(II) mediated phenol ring nitration by nitrogen dioxide.
Dalton transactions (Cambridge, England : 2003) (2015-10-31)
Vikash Kumar, Somnath Ghosh, Anoop Kumar Saini, Shaikh M Mobin, Biplab Mondal
ABSTRACT
Cu(II) complexes of N2O2 type ligands, L(1)H2 and L(2)H2 [L(1)H2 = 6,6'-(((pyridin-2-ylmethyl)azanediyl)bis(methylene))bis(2,4-di-tert-butylphenol); L(2)H2 = 2,4-di-tert-butyl-6-(((3-(tert-butyl)-2-hydroxy-5-methylbenzyl)(pyridin-2-yl-methyl)amino)methyl)phenol], have been synthesized. Addition of nitrogen dioxide (NO2) in THF solutions of the complexes resulted in the nitration at the 4-position of a coordinated equatorial phenolate ring of the ligand frameworks. This nitration did not occur at the phenol ring which is axially coordinated to the metal center. Spectroscopic evidence suggests that the reaction proceeds through a phenoxyl radical complex formation.
Product Description
Supelco
Model 1000 oxygen trap, connections size 1/4 in.