Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

503088

Supelco

Supelpure-O Oxygen/Moisture Trap

volume 750 cc, 1/4 in. fitting

Synonym(s):

SUPELPURE-O, 750CC, 1/4IN FITTINGS

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$720.00

$720.00


Available to ship onApril 17, 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$720.00

About This Item

UNSPSC Code:
12352200

$720.00


Available to ship onApril 17, 2025Details


Request a Bulk Order

volume

750 cc

fitting

1/4 in. fitting

Looking for similar products? Visit Product Comparison Guide

General description

The oxygen-removing catalysts can reduce oxygen to less than 2 ppb when the level in the incoming gas does not exceed 10 ppm. Because the catalyst is coated on a Molecular Sieve, this trap also can remove water vapor.

Dimensions:
  • 120 cc Trap: 111/8in. (28.2 cm) long x 13/8 in. (35 mm) diameter
  • 750 cc Trap: 161/2 in. (41.9 cm) long x 25/16in. (59 mm) diameter
  • S-Trap: 71/2 in. (19 cm) long x 59/16 in. (14.1 cm) wide x 1/2 in. (13 mm) diameter [total bed length = 193/4 in. (50.2 cm); the extended bed length ensures prolonged contact between the gas and the adsorbent and provides greater working capacity]

Application

Supelpure-O Oxygen/Moisture Trap along with Molecular Sieve 5A Moisture Trap was used as the two cascade columns required for the purification of ethylene, in an experimental study based on using micrometer-sized aggregates of polystyrene nanoparticles as fragmentable support for Ziegler-Natta catalysts.[1]

Legal Information

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic nanoparticles as fragmentable support for Ziegler-Natta catalysts.
Nietzel S
Journal of Polymer Science Part A: Polymer Chemistry, 53 (1), 15-22 (2015)

Questions

1–3 of 3 Questions  
  1. Is it possible to regenerate a spent trap through heat treatment of other method?

    1 answer
    1. No regeneration method is available for this product.

      Helpful?

  2. What is the pressure rating of the Supelpure _ O oxuygen/moisture trap, and what is the H2O/O2 capacities? Can it be used with H2?

    1 answer
    1. The maximum pressure recommended for this product is 250 psi (17.2 bar). The removal capacities are 10.2 grams of water and 2.4 grams of oxygen. This trap is suitable for use with hydrogen.

      Helpful?

  3. Does the "CC" designation relate to the volume of gas that the purifier can scrub at a time, or does it indicate the longevity of the purifier? Additionally, does it affect the flow rate? What pressure should a nitrogen regulator be set to in order to use this product?

    1 answer
    1. The "cc" designation refers to the volume of adsorbent material in each of the two products, Supelpure-O 120CC and Supelpure-O 750cc. The lifetime of the Supelpure-O, and/or any gas purification trap, relies on the contaminants and their concentration level in the incoming gas, as well as the flow rate. Bulletin 918 on gas purification includes a table of the maximum flow rate and pressure of various gas purification products, some of which are no longer available.
      The pressure setting on the incoming carrier gas tank depends on the type of analysis being performed, whether packed or capillary, isothermal or temperature programmed method, etc. For GC applications, a pressure at least 20psi higher than that achieved during the analysis is recommended, preferably 40-50psi higher. In GCs that do not maintain constant pressure, as the temperature increases, resistance and back pressure rise, causing the flow rate to drop.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service