Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z273252

Sigma-Aldrich

Detojet® low foaming liquid detergent

pkg of 947 mL in 1EA

Synonym(s):

USDA authorized cleaner, alkaline detergent, emulsifying detergent, low foam cleaner, machine washing detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

packaging

pkg of 947 mL in 1EA

manufacturer/tradename

Alconox, Inc. 1632

Looking for similar products? Visit Product Comparison Guide

General description

Det-o-jet is a low foaming, heavy duty alkaline detergent for use in labware washers, parts washers, power-spray systems, and ultrasonic cleaning systems. Concentrated, highly emulsifying and penetrating formula is free rinsing, and leaves no interfering residues. Protects interior working parts of washers. USDA authorized. Dilute 1:200. pH 12.4

  • Concentrated to save you money
  • Biodegradable and disposable
  • Low foaming heavy duty detergent for high performance results from your cleaning machinery
  • Free rinsing to give you reliable results and no interfering residues

Used to clean: Dissection instruments, laboratory ware, optical components, manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors. Authorized by USDA for use in federally inspected meat and poultry plants. FDA certified.

Used to remove: Soil, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, particulates, inorganic residues, chemical and solvents.

Surfaces cleaned: Recommended for glass, stainless steel, ceramic, cement and alkali resistant plastics and metals such as stainless steel. Corrosion testing is advisable.

Cleaning method: Laboratory warewashers, dishwashers, cart washers, spray washers, electrocleaning, ultrasonic cleaners flow through CIP (clean-in-place) and manual cleaning if proper skin and eye protection is used.

Directions: Follow machine manufacturers directions. Not intended for powder detergent receptacles. If available, use liquid detergent reservoir, connecting tubes or metering pump. In general, use 1/2 – 1% solution in hot water wash cycle (1 1/4 – 2 1/2 Tbsp. per gal., 3/4 – 1 1/4 oz. per gal. or 5-10 ml per liter—most machines need 2 1/2 oz. added at a typical 2 gal. wash cycle). For soak, circulate or ultrasonic cleaner use, make a 1% solution in cold, warm or hot water. For difficult soils, raise water temperature and use more detergent. Wear protective gloves and eyewear when handling.
RINSE THOROUGHLY—preferably with running water.
For critical cleaning, do final or all rinsing in distilled, deionized or purified water.
For food contact surfaces, rinse with potable water.

Legal Information

Detojet is a registered trademark of Alconox, Inc.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service