Key Documents
Z179981
Supelco
Michel-Miller column filter disc
Ace-Thred for #7
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
material
paper
manufacturer/tradename
Ace Glass 581406
technique(s)
LPLC: suitable
fitting
Ace-Thred for #7
General description
Use at either end of column. Filter disc sizes 7 and 11 are made from specially pure, very uniform, highly-absorbent paper. Order adapter separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)