Documents
Z503878
Scienceware® trays
w/o faucet, inside dimensions L × W × H 55 cm × 65 cm × 10.2 cm
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
material
low-density polyethylene container
manufacturer/tradename
Bel-Art
General description
Molded LDPE can withstand temperatures up to 77°C (171°F). The PE faucet has a 12.7 mm bore.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP