Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W282401

Sigma-Aldrich

Orange oil

sweet, Certified organic (NOP)

Synonym(s):

Citrus aurantium, Citrus sinensis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2824
MDL number:
UNSPSC Code:
12164502
grade:
Certified organic (NOP)
Halal
Kosher
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

Certified organic (NOP)
Halal
Kosher

origin

Costa Rica origin

refractive index

n20/D 1.473

density

0.844 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

orange

Looking for similar products? Visit Product Comparison Guide

Legal Information

Certified organic by MOSA

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean J Pendleton et al.
Journal of food science, 77(6), M308-M311 (2012-05-16)
Plant essential oils have previously been shown to exhibit antimicrobial activities against various microorganisms. In this study, cold pressed terpeneless Valencia orange oil (CPTVO) was examined at various temperatures (37, 10, and 4 °C) to determine its antimicrobial activity against
Misae Shinomiya et al.
Acta medica Okayama, 66(2), 111-118 (2012-04-25)
We studied the effects of natural essential oil on neurite outgrowth in PC12m3 neuronal cells to elucidate the mechanism underlying the action of the oils used in aromatherapy. Neurite outgrowth can be induced by nerve growth factor (NGF), where ERK
Mário Tanomaru-Filho et al.
Brazilian dental journal, 21(1), 46-49 (2010-05-14)
The aim of this study was to evaluate the effectiveness of 3 solvents (eucalyptol, orange oil, and xylol) on 2 types of gutta-percha (conventional and thermoplastic) and Resilon. Specimens (10 mm diameter x 1 mm thick; n=7 per condition) were
Jong-Hwan Lim et al.
Experimental animals, 61(1), 71-75 (2012-02-02)
This study was conducted to evaluate the oral absorption of enrofloxacin (ENFX) in rats when administered with orange oil or its main component, limonene. Compared with the group administered ENFX alone, the ENFX + limonene group did not show any
Kristina Kejlová et al.
Toxicology in vitro : an international journal published in association with BIBRA, 24(8), 2084-2089 (2010-08-07)
The aim of this study, linked-up with a previous study on bergamot oils, was the evaluation of phototoxic potential of essential oils (orange, lemon and Litsea cubeba), used as cosmetic ingredients. The applied tiered testing strategy included chemical analysis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service