Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

W265722

Sigma-Aldrich

Mandarin oil

greener alternative

natural, distilled, product of Simone Gatto

Sign Into View Organizational & Contract Pricing

Select a Size

W265722-SAMPLE-K
$214.00

$214.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W265722-SAMPLE-K
$214.00

About This Item

CAS Number:
FEMA Number:
2657
MDL number:
UNSPSC Code:
12164502
grade:
Kosher
natural
biological source:
Citrus reticulata Blanco (Citrus Nobilis (Mandarin Orange) Peel Oil)
food allergen:
no known allergens

$214.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

biological source

Citrus reticulata Blanco (Citrus Nobilis (Mandarin Orange) Peel Oil)

Quality Level

grade

Kosher
natural

reg. compliance

FDA 21 CFR 117

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

178 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

orange; floral; citrus

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

133.0 °F - closed cup

Flash Point(C)

56.11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service