Documents
TEM-LC200CU100
Sigma-Aldrich
Lacey Carbon Supported Copper Grids
size 200 mesh, size 100 nm , hole size, box of 25 ×
Synonym(s):
Lacey/ Carbon Film - Cu, 200 mesh 100 micron, 25pk
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25 ×
size
100 nm , hole size
200 mesh
General description
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.