Key Documents
930350
Sigma-Aldrich
TEM Grids
Carbon Film Supported Copper Hexagonal Mesh Grid, grid size 200 mesh, box of 50
Synonym(s):
TEM grids
Select a Size
1 EA
$169.00
$169.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$169.00
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
$169.00
Please contact Customer Service for Availability
Request a Bulk Order
Product Name
Carbon Film Supported Copper Hexagonal Mesh Grid, grid size 200 mesh, box of 50
material
carbon film
copper grid
description
film layer thickness 5 - 6 nm carbon
packaging
box of 50
grid size
200 mesh
General description
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 200 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Choose from one of the most recent versions:
Certificates of Analysis (COA)