Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

931519

Sigma-Aldrich

TEM Grids

Carbon Film Supported Copper Grid, Standard Thickness, grid size 300 mesh, box of 50

Synonym(s):

TEM grids

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$247.00

$247.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$247.00

About This Item

UNSPSC Code:
41111761
NACRES:
MA.24

$247.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Carbon Film Supported Copper Grid, Standard Thickness, grid size 300 mesh, box of 50

material

carbon film
copper grid

packaging

box of 50

film size

5-6 nm

grid size

300 mesh

Looking for similar products? Visit Product Comparison Guide

General description

TEM grids supported by carbon film only. A 5-6 nm thin film of pure carbon deposited on one side of the standard square mesh grid.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service