Key Documents
TEM-CFT200CU
Sigma-Aldrich
Carbon Film Supported Copper Thin Bar Square Mesh Grid
size 200 mesh, box of 25
Synonym(s):
CARBON FILM ONLY ON THIN BAR 200 MESH COPPER GRIDS 25/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-CFT200CU is not currently sold in your country. Contact Technical Service
packaging
box of 25
size
200 mesh
General description
A 5-6 nm thin film of pure carbon supported thin bar square mesh grid. Copper grid, 200 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Choose from one of the most recent versions:
Certificates of Analysis (COA)