Documents
CLS2480500
Pyrex® Allihn condenser, with male and female joints
jacket L 500 mm, joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint: ST/NS24/40
manufacturer/tradename
Corning 2480-500
jacket L
500 mm
Related Categories
General description
Suitable for use in vacuum distillation. With drip-tip
Legal Information
PYREX is a registered trademark of Corning, Inc.