Key Documents
SYNC244500
Synthware Allihn reflux condenser
joint: ST/NS 45/50, jacket L 260 mm
Synonym(s):
Synthware C244500, Soxhlet extraction condenser
Select a Size
1 EA
$171.00
$171.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$171.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$171.00
Please contact Customer Service for Availability
Joint
joint: ST/NS45/50
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C244500
jacket L
260 mm
General description
Allihn reflux condenser with a water cooled inner drip joint at bottom.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)