Skip to Content
MilliporeSigma
All Photos(1)

Documents

900862

Sigma-Aldrich

(4-(3-(Trifluoromethyl)-3H-diazirin-3-yl)-2-((trimethylsilyl)ethynyl)phenyl)methanol

Synonym(s):

Hydroxyl diazirine TMS-alkyne, Probe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15F3N2OSi
CAS Number:
Molecular Weight:
312.36
UNSPSC Code:
12352200
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

storage temp.

−20°C

SMILES string

OCC1=CC=C(C2(N=N2)C(F)(F)F)C=C1C#C[Si](C)(C)C

Application

(4-(3-(Trifluoromethyl)-3H-diazirin-3-yl)-2-((trimethylsilyl)ethynyl)phenyl)methanol is used for chemical probe synthesis, this trifunctional building block contains a light-activated diazirine, alkyne tag, and hydroxyl synthetic handle. When appended to a ligand or pharmacophore through its hydroxyl linker, this building block allows for UV light-induced covalent modification of a biological target with potential for downstream applications via the alkyne tag. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In partnership with Pfizer chemists, we have compiled a collection of trifunctional building blocks to enable development of probes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service