Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900616

Sigma-Aldrich

3-(But-3-yn-1-yl)-3-(2-iodoethyl)-3H-diazirine

≥95%

Synonym(s):

Iodo diazirine alkyne, Probe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9IN2
CAS Number:
Molecular Weight:
248.06
MDL number:
UNSPSC Code:
12352200

Assay

≥95%

form

liquid

reaction suitability

reaction type: click chemistry

storage temp.

−20°C

Application

Used for chemical probe synthesis, this trifunctional building block contains a light-activated diazirine, alkyne tag, and iodine synthetic handle. When appended to a ligand or pharmacophore through its iodinated linker, this building block allows for UV light-induced covalent modification of a biological target with potential for downstream applications via the alkyne tag. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service