Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900600

Sigma-Aldrich

3-(Fluorosulfonyl)-5-((trimethylsilyl)ethynyl)benzoic acid

≥95%

Synonym(s):

Carboxyl sulfonyl fluoride alkyne, Probe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13FO4SSi
CAS Number:
Molecular Weight:
300.38
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Assay

≥95%

form

solid

reaction suitability

reagent type: cross-linking reagent

availability

available only in USA

storage temp.

−20°C

SMILES string

O=S(C1=CC(C#C[Si](C)(C)C)=CC(C(O)=O)=C1)(F)=O

InChI key

CNXMDAYMSVWYAS-UHFFFAOYSA-N

Application

3-(Fluorosulfonyl)-5-((trimethylsilyl)ethynyl)benzoic acid is used for chemical probe synthesis, this trifunctional building block contains an aryl sulfonyl fluoride group, alkyne tag, and carboxylic acid synthetic handle. When appended to a ligand or pharmacophore through its acid linker, this building block allows for SuFEx-enabled, context-specific covalent modification of a biological target with potential for downstream applications via the alkyne tag. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Self-react. C - Skin Irrit. 2

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service