Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

464686

Sigma-Aldrich

Palladium(II) oxide

98%

Synonym(s):

Palladium monoxide, Palladium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
OPd
CAS Number:
Molecular Weight:
122.42
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

98%

mp

870 °C (lit.)

density

8.7 g/mL at 25 °C (lit.)

SMILES string

O=[Pd]

InChI

1S/O.Pd

InChI key

HBEQXAKJSGXAIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A new AB2O4 phase, LuPd2O4, with potential uses in superconducting material as well as being of goechemical importance, was formed at elevated pressure and temperature from Lu2O3 and PdO in the presence of potassium chlorate.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masanao Yokohira et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 58(4), 211-221 (2006-11-25)
We have established and documented an in vivo bioassay for detection of hazards with intratracheally instilled fine particles, which can be used for risk assessment of toxicity of materials inhaled into deep lung tissue of humans (Yokohira et al. Establishment
Self-assembly of a heteropolyoxopalladate nanocube: [Pd(II)13As(V)8O34(OH)6]8-.
Elena V Chubarova et al.
Angewandte Chemie (International ed. in English), 47(49), 9542-9546 (2008-10-14)
Qi Li et al.
Environmental science & technology, 44(9), 3493-3499 (2010-04-15)
A novel composite photocatalyst of nitrogen (N) and fluorine (F) codoped titanium oxide (TiO(2)) nanotube arrays with dispersed palladium oxide (PdO) nanoparticles was developed by dispersing PdO nanoparticles into N and F codoped TiO(2) nanotube array template created by anodization
Qi Li et al.
Environmental science & technology, 43(5), 1534-1539 (2009-04-09)
Removal of arsenic species from water by palladium-modified nitrogen-doped titanium oxide (TiON/PdO) nanoparticles was investigated with and without visible light. For the first time, a high degree of As(III) removal undervisible light illumination was demonstrated on oxide photocatalysts. Over 2
Matt W Ross et al.
The journal of physical chemistry. A, 117(6), 1030-1034 (2012-05-09)
Reported herein are strong-field ionization studies of small, neutral Pd(x)O(y) and Zr(x)O(y) clusters made using ultrafast laser pulses (~100 fs) centered at 624 nm. An enhancement in ionization of nearly 1.5 orders of magnitude lower in laser intensity than predicted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service