![Palladium(II) oxide (85% Pd) hydrogenation catalyst for synthesis](/deepweb/assets/sigmaaldrich/product/structures/252/621/570c8535-fe50-49fa-a170-3092f49d513c/640/570c8535-fe50-49fa-a170-3092f49d513c.png)
Palladium(II) oxide
![Palladium(II) oxide (85% Pd) hydrogenation catalyst for synthesis](/deepweb/assets/sigmaaldrich/product/structures/252/621/570c8535-fe50-49fa-a170-3092f49d513c/640/570c8535-fe50-49fa-a170-3092f49d513c.png)
Synonym(s):
Palladium(II) oxide (85% Pd)
Empirical Formula (Hill Notation):
OPd
CAS Number:
Molecular Weight:
122.42
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Reaction Type
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing