MilliporeSigma
  • Extreme ionization leading to Coulomb explosion of small palladium and zirconium oxide clusters and reactivity with carbon monoxide.
Extreme ionization leading to Coulomb explosion of small palladium and zirconium oxide clusters and reactivity with carbon monoxide.
The journal of physical chemistry. A (2012-05-09)
Matt W Ross, A W Castleman
ABSTRACT
Reported herein are strong-field ionization studies of small, neutral Pd(x)O(y) and Zr(x)O(y) clusters made using ultrafast laser pulses (~100 fs) centered at 624 nm. An enhancement in ionization of nearly 1.5 orders of magnitude lower in laser intensity than predicted from literature values is observed for both systems due to clustering. The change in enhancement upon addition of carbon monoxide at different pressures was also studied. Enhancement of high charge states of palladium was found to decrease upon CO addition, whereas in the case of the zirconium system, high charge states of zirconium were observed to increase. Pd and ZrO showed similar reactivity trends with CO and were found to have similar reactivity ratios in accord with their isovalent nature.
Sigma-Aldrich
Palladium(II) oxide, 99.97% trace metals basis