Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

403091

Sigma-Aldrich

Vinyl benzoate

≥99%, contains <=20 ppm Hydroquinone and/or <=50 ppm MEHQ as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH=CH2
CAS Number:
Molecular Weight:
148.16
Beilstein:
2041125
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.4 mmHg ( 25 °C)

Assay

≥99%

form

liquid

refractive index

n20/D 1.529 (lit.)

bp

95-96 °C/20 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=COC(=O)c1ccccc1

InChI

1S/C9H8O2/c1-2-11-9(10)8-6-4-3-5-7-8/h2-7H,1H2

InChI key

KOZCZZVUFDCZGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priyanka V Jawale et al.
Bioprocess and biosystems engineering, 44(2), 369-378 (2020-10-01)
The present study aimed to analyze reaction kinetics and mechanism for the synthesis of propyl benzoate in solvent-free conditions. Lipase was immobilized on Hydroxypropyl methylcellulose (HPMC) and polyvinyl alcohol (PVA) polymer blend by entrapment method. Among different lipases immobilized on
Shouwan Tang et al.
Chirality, 29(9), 512-521 (2017-06-22)
Six novel regioselectively substituted amylose derivatives with a benzoate at 2-position and two different phenylcarbamates at 3- and 6-positions were synthesized and their structures were characterized by
Rongling Yang et al.
Frontiers in bioengineering and biotechnology, 8, 273-273 (2020-05-05)
Highly efficient and regioselective synthesis of pharmacologically interesting aromatic esters of arbutin catalyzed by immobilized lipase from Penicillium expansum in co-solvent systems was successfully carried out. As compared to tetrahydrofuran solvent, the initial rate and substrate conversion of arbutin vanilylation
Eunae Cho et al.
Scientific reports, 6, 23740-23740 (2016-03-30)
Amphiphilic cyclodextrins have been synthesized with self-acylating reaction using vinyl esters in dimethylformamide. In the present study no base, catalyst, or enzyme was used, and the structural analyses using thin layer chromatography, nuclear magnetic resonance spectroscopy and mass spectrometry show
Renata Kołodziejska et al.
Chirality, 30(2), 206-214 (2017-11-16)
The application of ionic liquids as solvents for transesterification of prochiral pirymidine acyclonucleoside using lipase (EC 3.1.1.3) Amano PS from Burkholderia cepacia (BCL) is reported. The effect of using medium reaction, acyl group donor, and temperature on the activity and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service