MilliporeSigma
  • Preparation and evaluation of regioselectively substituted amylose derivatives for chiral separations.
Preparation and evaluation of regioselectively substituted amylose derivatives for chiral separations.
Chirality (2017-06-22)
Shouwan Tang, Zhaolei Jin, Baishen Sun, Fang Wang, Wenyuan Tang
ABSTRACT
Six novel regioselectively substituted amylose derivatives with a benzoate at 2-position and two different phenylcarbamates at 3- and 6-positions were synthesized and their structures were characterized by