Skip to Content
MilliporeSigma
All Photos(1)

Documents

380806

Sigma-Aldrich

Vinyl 4-tert-butylbenzoate

99%

Synonym(s):

Vinyl 4-(1,1-dimethylethyl)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4CO2CH=CH2
CAS Number:
Molecular Weight:
204.26
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

refractive index

n20/D 1.518 (lit.)

bp

100 °C/0.5 mmHg (lit.)

density

0.999 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)c1ccc(cc1)C(=O)OC=C

InChI

1S/C13H16O2/c1-5-15-12(14)10-6-8-11(9-7-10)13(2,3)4/h5-9H,1H2,2-4H3

InChI key

ZLHVSEPPILCZHH-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jérémie Brand et al.
Carbohydrate polymers, 169, 189-197 (2017-05-16)
Herein we propose a versatile method for the surface tailoring of cellulose nanocrystals (CNCs) based on the reactivity of vinyl ester molecules toward the accessible hydroxyl groups located at the surface of the nanoparticles. CNCs produced from wood pulp were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service