Skip to Content
MilliporeSigma
All Photos(1)

Documents

381551

Sigma-Aldrich

7-Isopropoxy-3-phenyl-4H-1-benzopyran-4-one

97%

Synonym(s):

Ipriflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16O3
CAS Number:
Molecular Weight:
280.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

116-120 °C (lit.)

SMILES string

CC(C)Oc1ccc2c(OC=C(C2=O)c3ccccc3)c1

InChI

1S/C18H16O3/c1-12(2)21-14-8-9-15-17(10-14)20-11-16(18(15)19)13-6-4-3-5-7-13/h3-12H,1-2H3

InChI key

SFBODOKJTYAUCM-UHFFFAOYSA-N

Gene Information

rat ... Alpl(25586)

Looking for similar products? Visit Product Comparison Guide

General description

7-Isopropoxy-3-phenyl-4H-1-benzopyran-4-one (Ipriflavone), a synthetic flavonoid, is reported to stimulate the activity of osteoblasts. It is reported to promote the deposition of calcium and the formation of mineralized nodules by newborn rat calvarial osteoblast-like (ROB) cells as well as the activity of alkaline phosphatase. Ipriflavone, an isoflavone derivative, is a new drug used to decrease bone loss in osteoporosis.

Application

7-Isopropoxy-3-phenyl-4H-1-benzopyran-4-one (Ipriflavone) has been used as a model drug in a study to functionalize the mesoporous bioactive glasses (MBG). Study suggested that since ipriflavone is a hydrophobic anti-osteoporotic drug, it easily attaches to the surface of MBG and results in long-term drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye J Chung et al.
Biopharmaceutics & drug disposition, 30(6), 294-304 (2009-07-11)
Ipriflavone, a derivative of naturally occurring isoflavones, was primarily metabolized in rats via hepatic CYP1A1/2 and 2C11. Protein and mRNA expression of CYP1A2 in the liver, reported to be increased in mutant Nagase analbuminemic rats (NARs), should influence the pharmacokinetic
Shirin Hooshmand et al.
Clinical nutrition (Edinburgh, Scotland), 27(4), 643-648 (2008-06-24)
Recent reports have indicated that soy isoflavones may be protective against breast cancer. However, the effects of the synthetic isoflavone, ipriflavone, on mammary tumorigenisis, alone or in combination with genistin, a soy isoflavone, have not been investigated. Eighty-eight 36-day-old female
Jie Wu et al.
Climacteric : the journal of the International Menopause Society, 11(3), 212-220 (2008-06-24)
To evaluate the effects of estrogen and estrogenic compounds on cognition in ovariectomized rats. Female Sprague-Dawley rats (3-5 months old) weighing 250-300 g were randomly divided into seven groups: Sham, ovariectomized (OVX), OVX plus estradiol valerate, OVX plus ipriflavone, OVX
[Effect of ipriflavone on growth and blood testosterone level in male rats].
Hui-jun Guo et al.
Zhongguo ying yong sheng li xue za zhi = Zhongguo yingyong shenglixue zazhi = Chinese journal of applied physiology, 25(2), 170-171 (2009-05-01)
Hanaa S El-Desoky et al.
Journal of AOAC International, 92(3), 806-812 (2009-07-21)
Ipriflavone is a nonhormonal isoflavone derivative currently used as a bone-building agent for prevention and treatment of postmenopausal osteoporosis. Ipriflavone was reduced at the hanging mercury drop electrode in buffers of pH 2-11 via the consumption of 2 electrons corresponding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service