Skip to Content
MilliporeSigma
7-Isopropoxy-3-phenyl-4H-1-benzopyran-4-one 97%

7-Isopropoxy-3-phenyl-4H-1-benzopyran-4-one

7-Isopropoxy-3-phenyl-4H-1-benzopyran-4-one 97%
Synonym(s):
Ipriflavone
Empirical Formula (Hill Notation):
C18H16O3
CAS Number:
Molecular Weight:
280.32

Products

Product Category
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing